missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SIX2 Polyclonal Antibody

Product Code. 17227733 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17227733 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17227733 Supplier Invitrogen™ Supplier No. PA5114967
  • ONLINE EXCLUSIVE PRICE
    506.00 €
    359.26 € / 100µL
    Save 146.74 €
    29% Off
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SIX2.

This gene is a member of the vertebrate gene family which encode proteins homologous to the Drosophila 'sine oculis' homeobox protein. SIX2 is a transcription factor which, like other members of this gene family, may be involved in limb or eye development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIX2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene SIX2
Gene Accession No. Q62232, Q9NPC8
Gene Alias homeobox protein SIX2; Sine oculis homeobox homolog 2; sine oculis-related homeobox 2; sine oculis-related homeobox 2 homolog; SIX homeobox 2; SIX2
Gene Symbols SIX2
Host Species Rabbit
Immunogen A synthesized peptide derived from human SIX2(Accession Q9NPC8), corresponding to amino acid residues S207-V257.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10736, 20472, 366542
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.