missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SKAP Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93210-0.1ml
This item is not returnable.
View return policy
Description
SKAP Polyclonal antibody specifically detects SKAP in Human samples. It is validated for Western Blot
Specifications
| SKAP | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| C15orf23, chromosome 15 open reading frame 23, FLJ14502, HSD11, MGC141728, MGC141729, putative TRAF4-associated factor 1, SKAP, small kinetochore associated protein, TRAF4 associated factor 1, TRAF4AF1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human KNSTRN (NP_001136233.1). MAAPEAPPLDRVFRTTWLSTECDSHPLPPSYRKFLFETQAADLAGGTTVAAGNLLNESEKDCGQDRRAPGVQPCRLVTMTSVVKTVYSLQPPSALSGGQPADTQTRATSKSLLPVRSKEVDVSKQLHSGGPENDVTKITKLRRENGQMKATDTATRRNVRKGYKPLSKQKSEEELKDKNQLLEAVNKQLHQKLTETQGELKDLTQKVELLEKFRDNCLAILESKGLDPALGSETLASRQESTTDHMDSMLLLETLQEELKLFNETAKKQM | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 90417 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction