missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SKOR2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP2-14565
This item is not returnable.
View return policy
Description
SKOR2 Polyclonal specifically detects SKOR2 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| SKOR2 | |
| Polyclonal | |
| Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| CORL2, FUSSEL18, SKOR2 SKI family transcriptional corepressor 2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 652991 | |
| Human, Mouse | |
| IgG |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| SKOR2 | |
| This antibody was developed against a recombinant protein corresponding to the amino acids: MASSPLPGPNDILLASPSSAFQPDTLSQPRPGHANLKPNQVGQVILYGIPIVS | |
| 0.1 mL | |
| Primary | |
| Specificity of human SKOR2 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction