missing translation for 'onlineSavingsMsg'
Learn More

SL 0101-1, MedChemExpress

Product Code. 16461489
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16461489 1 mg 1mg
16428977 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16461489 Supplier Medchem Express Supplier No. HY15237
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress SL 0101-1 (SL0101), a kaempferol glycoside, isolated from the tropical plant F. refracta, is a cell-permeable, selective, reversible, ATP-competitive p90 Ribosomal S6 Kinase (RSK) inhibitor, with an IC50 of 89 nM. SL 0101-1 (SL0101) is a selective RSK1/2 inhibitor, with a Ki of 1 μM.

SL 0101-1 (SL0101), a kaempferol glycoside, isolated from the tropical plant F. refracta, is a cell-permeable, selective, reversible, ATP-competitive p90 Ribosomal S6 Kinase (RSK) inhibitor, with an IC50 of 89 nM. SL 0101-1 (SL0101) is a selective RSK1/2 inhibitor, with a Ki of 1 μM.

Specifications

Chemical Name or Material SL 0101-1
CAS 77307-50-7
Color White
Purity Grade Notes Research
Molecular Formula C25H24O12
Quantity 1 mg
Synonym SL0101
SMILES OC1=CC(O)=C(C(C(O[C@@H]2O[C@@H](C)[C@H](OC(C)=O)[C@@H](OC(C)=O)[C@H]2O)=C(C3=CC=C(O)C=C3)O4)=O)C4=C1
Molecular Weight (g/mol) 516.45
Formula Weight 516.45
Percent Purity 96.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.