missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ SLAM/CD150 Antibody Pair [HRP]

Product Code. 18202446 Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
This item is not returnable. View return policy

Product Code. 18202446

Brand: Novus Biologicals™ NBP27930715PLATES

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human SLAM/CD150.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 7.8125 pg/mL
Assay Sensitivity 7.81-500 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human SLAM/CD150.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias CD 150, CD150IPO-3, CDw150, signaling lymphocytic activation molecule, signaling lymphocytic activation molecule family member 1, SLAMCD150 antigen
Gene ID (Entrez) 6504
Gene Symbol SLAMF1
Host Species Mouse/Rabbit
Kit Contents
  • Detection Antibody: 0.2 mg/mL mouse anti-SLAM/CD150 monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4).
  • Capture Antibody: 1.0 mg/mL of rabbit anti-SLAM/CD150 monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 33 ng of recombinant SLAM/CD150.
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Cell Cycle and Replication, Hematopoietic Stem Cell Markers, Phospho Specific, Stem Cell Markers
    Storage Requirements Storage is content dependent.
    Target SLAM/CD150
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 0.5 ng/mL is recommended.
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.