missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC12A9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
415.00 € - 589.00 €
Specifications
| Antigen | SLC12A9 |
|---|---|
| Dilution | Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18625428
|
Novus Biologicals
NBP2-68858-25ul |
25 μL |
415.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18611519
|
Novus Biologicals
NBP2-68858 |
100 μg |
589.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SLC12A9 Polyclonal antibody specifically detects SLC12A9 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| SLC12A9 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| solute carrier family 12 member 9, CCC6, CIP1, hCCC6, solute carrier family 12 (potassium/chloride transporters), member 9, WO3.3 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: AWHSARLRIFLCLGPREAPGAAEGRLRALLSQLRIRAEVQEVVWGEGAGAGEPEAEEEGDFVNSGRGDAEAEALAR | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol | |
| 56996 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title