missing translation for 'onlineSavingsMsg'
Learn More

SLC16A3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 6 publications

280.00 € - 572.00 €

Specifications

Antigen SLC16A3
Dilution Western Blot Reported in scientific literature (PMID:33212210), Simple Western 1:250, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500, Immunohistochemistry-Frozen Reactivity reported in (PMID:24692237), Immunoblotting Reactivity reported in (PMID: 28978054).
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18489840
View Documents
Novus Biologicals
NBP1-81251-25ul
25ul
280.00 €
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18707913
View Documents
Novus Biologicals
NBP1-81251
572.00 €
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SLC16A3 Polyclonal specifically detects SLC16A3 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting.
Specifications

Specifications

SLC16A3
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot
Unconjugated
RUO
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
MCT 3, MCT3MCT 4, MCT4, MCT4MGC138472, MGC138474, Monocarboxylate transporter 3, monocarboxylate transporter 4, solute carrier family 16 (monocarboxylic acid transporters), member 3, Solute carrier family 16 member 3, solute carrier family 16, member 3 (monocarboxylic acid transporter 4)
SLC16A3
IgG
Affinity Purified
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Western Blot Reported in scientific literature (PMID:33212210), Simple Western 1:250, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500, Immunohistochemistry-Frozen Reactivity reported in (PMID:24692237), Immunoblotting Reactivity reported in (PMID: 28978054).
Polyclonal
Rabbit
Human
O15427
9123
This antibody was developed against Recombinant Protein corresponding to amino acids:AEEEKLHKPPADSGVDLREVEHFLKAEPEKNGEVVHTPETSV
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Certificates

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.