missing translation for 'onlineSavingsMsg'
Learn More

SLC18A1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17218693 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17218693 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17218693 Supplier Thermo Scientific Supplier No. PA5115909

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

SLC18A1 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC18A1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene SLC18A1
Gene Accession No. P54219, Q01818, Q8R090
Gene Alias CGAT; VMAT
Gene Symbols SLC18A1
Host Species Rabbit
Immunogen A synthesized peptide derived from human SLC18A1(Accession P54219), corresponding to amino acid residues R479-E525.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110877, 25693, 6570
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.