missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLC22A15 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15887683
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15887683 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15887683 Supplier Invitrogen™ Supplier No. PA569276

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

Organic ion transporters, such as SLC22A15, transport various medically and physiologically important compounds, including pharmaceuticals, toxins, hormones, neurotransmitters, and cellular metabolites. These transporters are also referred to as amphiphilic solute facilitators (ASFs).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC22A15
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC22A15
Gene Accession No. Q8IZD6
Gene Alias 2610034P21Rik; 4930477M19; A530052I06Rik; BB219042; flipt 1; FLIPT1; fly-like putative organic ion transporter 1; fly-like putative transporter 1; PRO34686; SLC22A15; solute carrier family 22 (organic anion/cation transporter), member 15; solute carrier family 22 (organic cation transporter), member 15; solute carrier family 22 member 15; solute carrier family 22, member 15; trans-like protein; UNQ9429/PRO34686
Gene Symbols SLC22A15
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human SLC22A15.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55356
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.