Learn More
SLC22A2/OCT2 Antibody (CL0628) [CoraFluor™ 1], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52941CL1
Product Code. 30289101
  • 1175.00 € / 0.10mL
Description
SLC22A2/OCT2 Monoclonal antibody specifically detects SLC22A2/OCT2 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SLC22A2/OCT2
Monoclonal
CoraFluor 1
hOCT2, MGC32628, OCT2Organic cation transporter 2, solute carrier family 22 (organic cation transporter), member 2, solute carrier family 22 member 2
This antibody was developed using a recombinant protein derived from O15244, with the exact immunogen sequence remaining proprietary.
0.1 mL
Primary
Human
Purified
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL0628
PBS
Mouse
Protein A purified
RUO
6582
Store at 4⪚C in the dark. Do not freeze.
IgG1