missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC23A1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-13318
This item is not returnable.
View return policy
Description
SLC23A1 Polyclonal specifically detects SLC23A1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| SLC23A1 | |
| Polyclonal | |
| Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| hSVCT1, SLC23A2, Sodium-dependent vitamin C transporter 1, sodium-dependent vitamin C transporter-1, solute carrier family 23 (nucleobase transporters), member 1, solute carrier family 23 (nucleobase transporters), member 2, solute carrier family 23 member 1, SVCT1Na(+)/L-ascorbic acid transporter 1, Yolk sac permease-like molecule 3, YSPL3MGC22361 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 9963 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| SLC23A1 | |
| This antibody was developed against a recombinant protein corresponding to the amino acids: NTVPGSPEERGLIQWKAGAHANSDMSSSLKSYDFPIGMGIVKRITFLKYIPICPVFKGFSSSSKDQIAIPEDTPENTETAS | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction