missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC25A12 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-89020
This item is not returnable.
View return policy
Description
SLC25A12 Polyclonal specifically detects SLC25A12 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| SLC25A12 | |
| Polyclonal | |
| Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20-1:50 | |
| O75746 | |
| SLC25A12 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:FNWDCEFIRLHFGHNRKKHLNYTEFTQFLQELQLEHARQAFALKDKSKSGMISGLDFSDIMV | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| AGC1, araceli hiperlarga, ARALAR, ARALAR1, calcium binding mitochondrial carrier superfamily member Aralar1, calcium-binding mitochondrial carrier protein Aralar1, Mitochondrial aspartate glutamate carrier 1, solute carrier family 25 (mitochondrial carrier, Aralar), member 12, Solute carrier family 25 member 12 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 8604 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction