missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC25A12 Recombinant Rabbit Monoclonal Antibody (24GB2340)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557714
This item is not returnable.
View return policy
Description
SLC25A12 Recombinant Monoclonal Antibody for Western Blot, Flow
This gene encodes a calcium-binding mitochondrial carrier protein. The encoded protein localizes to the mitochondria and is involved in the exchange of aspartate for glutamate across the inner mitochondrial membrane. Polymorphisms in the gene encoding this protein may be associated with risk of autism.
Specifications
| SLC25A12 | |
| Recombinant Monoclonal | |
| 0.8 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O75746, Q8BH59 | |
| SLC25A12 | |
| A synthesized peptide derived from human SLC25A12 (aa 601-650). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB2340 | |
| Unconjugated | |
| SLC25A12 | |
| 2610002D09Rik; AGC1; AI839531; araceli hiperlarga; ARALAR; Aralar1; B230107K20Rik; BB129864; calcium binding mitochondrial carrier superfamily member Aralar1; calcium-binding mitochondrial carrier protein Aralar1; Calcium-binding mitochondrial carrier protein Aralar1-like protein; mitochondrial aspartate glutamate carrier 1; RGD1561141; SLC25A12; solute carrier family 25 (aspartate/glutamate carrier), member 12; solute carrier family 25 (mitochondrial carrier, Aralar), member 12; solute carrier family 25 member 12; solute carrier family 25, member 12 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 362145, 78830, 8604 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction