missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC30A1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marke: Novus Biologicals NBP2-54987-25ul
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
SLC30A1 Polyclonal specifically detects SLC30A1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Spezifikation
| SLC30A1 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1 - 4 μg/mL | |
| solute carrier family 30 (zinc transporter), member 1, Solute carrier family 30 member 1, zinc transporter 1, ZnT-1, ZNT1znT-1, ZRC1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 7779 | |
| Human | |
| IgG |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| SLC30A1 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:RNVEGVEEVHELHVWQLAGSRIIATAHIKCEDPTSYMEVAKTIKDVFHNHGIHATTIQPEFASVGSKSSVVPCELACRTQCALKQCCGTLPQAPSGKDAEKTPAVSISCLELSNNLEKKPRRTKAENIPAVVIEIKNMPNKQPESS | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur