missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GM-CSF Antibody (BVD2-21C11), Janelia Fluor™ 646, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-34570JF646
This item is not returnable.
View return policy
Description
GM-CSF Monoclonal antibody specifically detects GM-CSF in Human, Cynomolgus Monkey, Macaque samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen), Neutralization, CyTOF
Specifications
| GM-CSF | |
| Monoclonal | |
| Janelia Fluor 646 | |
| colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, molgramostin, sargramostim | |
| Recombinant human GM-CSF protein (Uniprot: P04141) | |
| 0.1 mL | |
| Biologically Active Proteins, Cytokine Research, Immunology | |
| 1437 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation | |
| BVD2-21C11 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Cynomolgus Monkey, Macaque | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction