missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC35B3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
415.00 € - 624.00 €
Specifications
| Antigen | SLC35B3 |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18613946
|
Novus Biologicals
NBP2-37965-25ul |
25 μL |
415.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18178397
|
Novus Biologicals
NBP2-37965 |
0.1 mL |
624.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SLC35B3 Polyclonal specifically detects SLC35B3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| SLC35B3 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 3' phosphoadenosine 5' phosphosulfate transporter 2, adenosine 3'-phospho 5'-phosphosulfate transporter 2, C6orf196, CGI-19, chromosome 6 open reading frame 196, PAPS transporter 2, PAPST2dJ453H5.1,3'-phosphoadenosine 5'-phosphosulfate transporter, Solute carrier family 35 member B3, solute carrier family 35, member B3 | |
| SLC35B3 | |
| IgG | |
| Affinity Purified |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q9H1N7 | |
| 51000 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: QQAKDIQNITVQETNKNNSESIECSKITMDLKFNNSRKYISITVPSKTQTMSPHIKSVDDVVVLGMNL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title