missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC4A2 Polyclonal Antibody
Description
Recommended positive controls: A549. Predicted reactivity: Mouse (97%), Rat (96%), Rabbit (96%), Chicken (82%), Bovine (98%), Guinea pig (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Plasma membrane anion exchange protein of wide distribution.
Specifications
Specifications
| Antigen | SLC4A2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.74 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | SLC4A2 |
| Gene Accession No. | P04920 |
| Gene Alias | AE 2; AE2; Aep2; anion exchange protein 2; anion exchanger 2; anion exchanger 2 type a; anion exchanger 2 type b1; anion exchanger 2 type b2; B3RP; B3rp2; B3RP-2; Band 3-related protein; band 3-related protein 2; BND3L; EPB3L1; Erythrocyte band 3; erythrocyte membrane protein band 3-like 1; HKB3; MPB3L; NBND3; non-erythroid band 3-like protein; Slc4a2; solute carrier family 4 (anion exchanger), member 2; solute carrier family 4 member 2; Solute carrier family 4, member 2, anion exchange protein 2 |
| Gene Symbols | SLC4A2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?