missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC52A3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94430-0.02ml
This item is not returnable.
View return policy
Description
SLC52A3 Polyclonal antibody specifically detects SLC52A3 in Mouse samples. It is validated for Western Blot
Specifications
| SLC52A3 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| bA371L19.1, BVVLS, BVVLS1, C20orf54, hRFT2, RFT2, RFVT3, SLC52A3 solute carrier family 52, riboflavin transporter, member 3 | |
| A synthetic peptide corresponding to a sequence within amino acids 200-300 of human SLC52A3 (NP_212134.3). PGMEAPLSHLESRYLPAHFSPLVFFLLLSIMMACCLVAFFVLQRQPRCWEASVEDLLNDQVTLHSIRPREENDLGPAGTVDSSQGQGYLEEKAAPCCPAHL | |
| 0.02 mL | |
| Cancer, Endocrinology, Signal Transduction | |
| 113278 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction