missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC5A8 Monoclonal Antibody (903502)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561948
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
SLC5A8 has been shown to transport iodide by a passive mechanism (Rodriguez et al., 2002 ) and monocarboxylates and short-chain fatty acids by a sodium-coupled mechanism (Gopal et al., 2004 ).
Specifications
| SLC5A8 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q8N695 | |
| Slc5a8 | |
| NS0 mouse myeloma cell line transfected with human SLC5A8/SMCT1, Met1-Leu610. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry | |
| 903502 | |
| Unconjugated | |
| Slc5a8 | |
| AIT; Apical iodide transporter; apical iodide transporter , putative; electrogenic sodium monocarboxylate cotransporter; RGD1564146; Slc5a8; SMCT; SMCT1; Sodium iodide-related cotransporter; sodium-coupled monocarboxylate transporter 1; solute carrier family 5 (iodide transporter), member 8; solute carrier family 5 (sodium/monocarboxylate cotransporter), member 8; solute carrier family 5 member 8 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 160728 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction