missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC6A15 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-83867
This item is not returnable.
View return policy
Description
SLC6A15 Polyclonal specifically detects SLC6A15 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| SLC6A15 | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| B0AT2, DKFZp761I0921, FLJ10316, homolog of rat orphan transporter v7-3, hv7-3, MGC87066, NTT73orphan sodium- and chloride-dependent neurotransmitter transporter NTT73, orphan transporter v7-3, SBAT1, Sodium- and chloride-dependent neurotransmitter transporter NTT73, sodium/chloride dependent neurotransmitter transporter Homo sapiens orphanneurotransmitter transporter NTT7, Sodium-coupled branched-chain amino-acid transporter 1, solute carrier family 6 (neurotransmitter transporter), member 15, solute carrier family 6 (neutral amino acid transporter), member 15, Solute carrier family 6 member 15, solute carrier family 6, member 15, Transporter v7-3, V7-3 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 55117 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| SLC6A15 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:RELDDDVTESVKDLLSNEDAADDAFKTSELIVDGQEEKDTDVEEGSEVEDERPAWNSKLQ | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction