missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC6A3/DAT1 Antibody (CL3123), Novus Biologicals™
Mouse Monoclonal Antibody
302.00 € - 513.00 €
Specifications
| Antigen | SLC6A3/DAT1 |
|---|---|
| Clone | CL3123 |
| Dilution | Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18604436
|
Novus Biologicals
NBP2-46649-25ul |
25 μL |
302.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18626055
|
Novus Biologicals
NBP2-46649 |
0.1 mL |
513.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SLC6A3/DAT1 Monoclonal antibody specifically detects SLC6A3/DAT1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| SLC6A3/DAT1 | |
| Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Q01959 | |
| 6531 | |
| IgG1 | |
| Protein A purified |
| CL3123 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Neuroscience | |
| PBS (pH 7.2), 40% Glycerol | |
| DA transporter, DATDAT1Solute carrier family 6 member 3, sodium-dependent dopamine transporter, solute carrier family 6 (neurotransmitter transporter, dopamine), member 3 | |
| Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence LHYLFSSFTTELPWNNSWNSPNCSDAHPGDSSGDSSGLNDTFGTTPAAEYFERGVLHLHQSHGIDDLG | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title