missing translation for 'onlineSavingsMsg'
Learn More

SLC7A5/LAT1 Antibody (BU53) - BSA Free, R&D Systems™

Product Code. 18606736
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
Unit Size:
0.03mL
0.1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18606736 0.025 mL 0.03mL
18676108 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18606736 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP250465SS
  • ONLINE EXCLUSIVE PRICE
    251.68 €
    207.90 € / 0.03mL
    Save 43.78 €
    17% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

SLC7A5/LAT1 Monoclonal specifically detects SLC7A5/LAT1 in Human samples. It is validated for CyTOF, Flow Cytometry, Flow Cytometry (Cell Surface), Immunohistochemistry, Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC7A5/LAT1
Applications CyTOF, Flow Cytometry, Flow Cytometry (Cell Surface), Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone BU53
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1 - 2 μg/mL, Immunohistochemistry 1:100 - 1:300, Immunohistochemistry-Paraffin 1:100 - 1:300, Flow (Cell Surface) 1 - 2 μg/mL, CyTOF-ready
Formulation PBS
Gene Alias 4F2 LC, CD98, CD98LC, D16S469EL-type amino acid transporter 1, E16, Integral membrane protein E16, large neutral amino acids transporter 1, large neutral amino acids transporter small subunit 1, LAT1hLAT1, MPE16CD98 light chain, sodium-independent neutral amino acid transporter LAT1,4F2LC, solute carrier family 7 (cationic amino acid transporter, y+ system), member 5,4F2 light chain, Solute carrier family 7 member 5, y+ system cationic amino acid transporter
Host Species Mouse
Immunogen Plasma cell line 8226
Purification Method Protein G purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 8140
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG2a
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.