missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC9A9 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-95245-0.1ml
This item is not returnable.
View return policy
Description
SLC9A9 Polyclonal antibody specifically detects SLC9A9 in Human, Mouse samples. It is validated for Western Blot
Specifications
| SLC9A9 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| AUTS16, FLJ35613, Na(+)/H(+) exchanger 9, NHE9, NHE-9, putative protein product of Nbla00118, sodium/hydrogen exchanger 9, sodium/proton exchanger NHE9, solute carrier family 9 (sodium/hydrogen exchanger), solute carrier family 9 (sodium/hydrogen exchanger), isoform 9, solute carrier family 9 (sodium/hydrogen exchanger), member 9, Solute carrier family 9 member 9 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-130 of human SLC9A9 (NP_775924.1). MERQSRVMSEKDEYQFQHQGAVELLVFNFLLILTILTIWLFKNHRFRFLHETGGAMVYGLIMGLILRYATAPTDIESGTVYDCVKLTFSPSTLLVNITDQVYEYKYKREISQHNINPHQGNAILEKMTFD | |
| 0.1 mL | |
| Cancer, Endocrinology, Signal Transduction | |
| 285195 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction