missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLCO4A1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-80977-25ul
This item is not returnable.
View return policy
Description
SLCO4A1 Polyclonal specifically detects SLCO4A1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| SLCO4A1 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| colon organic anion transporter, OATP1, OATP4A1, OATPE, OATP-E, OATP-RP1, organic anion-transporting polypeptide E, POAT, SLC21A12, solute carrier family 21 (organic anion transporter), member 12, solute carrier family 21 member 12, solute carrier organic anion transporter family member 4A1, solute carrier organic anion transporter family, member 4A1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 28231 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| SLCO4A1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:GRYEVELDAGVRTCPANPGAVCADSTSGLSRYQL | |
| 25ul | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction