missing translation for 'onlineSavingsMsg'
Learn More

SLITRK3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

280.00 € - 624.00 €

Specifications

Antigen SLITRK3
Dilution Immunohistochemistry 1:2500 - 1:5000, Immunohistochemistry-Paraffin 1:2500 - 1:5000
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18693658
View Documents
Novus Biologicals
NBP2-62616-25ul
25 μL
280.00 €
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18628848
View Documents
Novus Biologicals
NBP2-62616
100 μg
624.00 €
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SLITRK3 Polyclonal antibody specifically detects SLITRK3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

SLITRK3
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Neuroscience
PBS (pH 7.2) and 40% Glycerol
22865
IgG
Protein A purified
Immunohistochemistry 1:2500 - 1:5000, Immunohistochemistry-Paraffin 1:2500 - 1:5000
Polyclonal
Purified
RUO
Human
MGC138681, SLIT and NTRK-like family, member 3
This antibody was developed against a recombinant protein corresponding to amino acids: EEVAVSSAQEAGSAERGGPGTQPPGMGEALLGSEQFAETPKENHSNYRTLLEKEKEWALAVSSSQLNTIVTVNHHHPHHPAVGGVSGVVGGTGGDLAGFRHHEKNGGVVLFPPGGGCGSGSMLLDRERPQPAPCTVGFVDCLY
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.