missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLITRK5 Monoclonal Antibody (310516)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561181
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
SLIT and NTRK-like family 5 (Slitrk5) is a member a protein family consisting of six homologous transmembrane proteins (Slitrk1-6) that share two conserved leucine-rich repeat domains in the extracellular domain and have significant homology to Slit, a secreted axonal growth-controlling protein. These proteins are also homologous to trk neurotrophin receptors in their intracellular domains. Expression of Slitrk proteins is highly restricted to neural and brain tumor tissues, but varies within the protein family. Like every other Slitrk protein except Slitrk1, overexpression of Slitrk5 inhibited neurite outgrowth in cultured neurons, suggesting that these proteins are involved in the control of neurite outgrowth.
Specifications
| SLITRK5 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| O94991 | |
| SLITRK5 | |
| Mouse myeloma cell line NS0-derived recombinant human SLITRK5, Glu39-Val665. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| 310516 | |
| Unconjugated | |
| SLITRK5 | |
| 2610019D03Rik; bA364G4.2; KIAA0918; leucine rich repeat containing 11; leucine-rich repeat-containing protein 11; LRRC11; SLIT and NTRK like family member 5; SLIT and NTRK-like family, member 5; SLIT and NTRK-like protein 5; slit and trk like gene 5; Slitrk5 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 26050 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction