missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SLITRK6 Antibody (sirtratumab) - IgG2SA, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28589-50ug
This item is not returnable.
View return policy
Description
SLITRK6 Monoclonal antibody specifically detects SLITRK6 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| SLITRK6 | |
| Monoclonal | |
| Unconjugated | |
| Q9H5Y7 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG2 |
| Flow Cytometry, ELISA, Functional Assay | |
| sirtratumab | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| FLJ22774, MGC119595, MGC119596, MGC119597,4832410J21Rik, SLIT and NTRK-like family, member 6, SLIT and NTRK-like protein 6, slit and trk like gene 6 | |
| SLITRK6 | |
| 50 μg | |
| Neuroscience | |
| 84189 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction