missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMAC/Diablo Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76772-0.025mg
This item is not returnable.
View return policy
Description
SMAC/Diablo Polyclonal antibody specifically detects SMAC/Diablo in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| SMAC/Diablo | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| 0610041G12Rik, diablo, IAP-binding mitochondrial protein, Direct IAP-binding protein with low pI, mitochondrial Smac protein, Second mitochondria-derived activator of caspase, Smac, SMACmitochondrial | |
| Antibody was raised against a 16 amino acid peptide near the carboxy terminus of murine Smac. The immunogen is located within the last 50 amino acids of Smac. Amino Acid Squence: SDEGADQEEEAYLRED | |
| 0.025 mg | |
| Apoptosis, Cancer, Cell Biology, Cellular Markers, Mitochondrial Markers, Tumor Suppressors | |
| 56616 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2 μg/mL, Immunocytochemistry/ Immunofluorescence 10 μg/mL, Immunoprecipitation, Immunohistochemistry-Paraffin | |
| AF203914 | |
| Rabbit | |
| Ion exchange chromatography | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction