missing translation for 'onlineSavingsMsg'
Learn More

SMAC/Diablo Antibody (S08-6A3), Novus Biologicals™

Rabbit Monoclonal Antibody

421.00 € - 645.00 €

Specifications

Antigen SMAC/Diablo
Clone S08-6A3
Concentration 0.3 mg/mL
Dilution Western Blot, Immunohistochemistry 1:20-1:100, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18647743
View Documents
Novus Biologicals
NBP3-19775-100ul
100 μg
645.00 €
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18608453
View Documents
Novus Biologicals
NBP3-19775-50ul
50 μg
421.00 €
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SMAC/Diablo Monoclonal antibody specifically detects SMAC/Diablo in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications

Specifications

SMAC/Diablo
0.3 mg/mL
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Apoptosis, Cancer, Cell Biology, Cellular Markers, Mitochondrial Markers, Tumor Suppressors
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
56616
IgG
Affinity purified
S08-6A3
Western Blot, Immunohistochemistry 1:20-1:100, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Monoclonal
Purified
RUO
Human, Mouse, Rat
0610041G12Rik, diablo, IAP-binding mitochondrial protein, Direct IAP-binding protein with low pI, mitochondrial Smac protein, Second mitochondria-derived activator of caspase, Smac, SMACmitochondrial
A synthetic peptide of human SMAC/Diablo (Uniprot # Q9NR28)
Primary
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.