missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZIC1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
Brand: Novus Biologicals NB600-488SS
This item is not returnable.
View return policy
Description
ZIC1 Polyclonal specifically detects ZIC1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| ZIC1 | |
| Polyclonal | |
| Unconjugated | |
| PBS and 30% Glycerol with 0.01% Sodium Azide | |
| Zic family member 1 (odd-paired homolog, Drosophila), ZICZic family member 1 (odd-paired Drosophila homolog), Zinc finger protein 201, Zinc finger protein of the cerebellum 1, ZNF201zinc finger protein ZIC 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human and mouse. Predicted to react with rat based on 100% sequence homology. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1:1000, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:400, Immunohistochemistry-Frozen reported in scientific literature (PMID 23862012) | |
| Q15915 | |
| ZIC1 | |
| A synthetic peptide made to an internal portion of the human Zic1 protein (between residues 20-75) [UniProt Q15915] | |
| 0.025 mL | |
| Growth and Development, Neuronal Cell Markers, Neuroscience, Signal Transduction | |
| 7545 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction