missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Smad2 Antibody (CL8472) [DyLight 650], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-88916C
This item is not returnable.
View return policy
Description
Smad2 Monoclonal antibody specifically detects Smad2 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| Smad2 | |
| Monoclonal | |
| DyLight 650 | |
| hMAD-2, hSMAD2, JV18, JV18-1SMAD, mothers against DPP homolog 2 (Drosophila), MAD homolog 2, Mad protein homolog, Mad, mothers against decapentaplegic homolog 2, MADH2MGC34440, MADR2MGC22139, Mad-related protein 2, mother against DPP homolog 2, mothers against decapentaplegic homolog 2, Mothers against DPP homolog 2, Sma- and Mad-related protein 2, SMAD 2, SMAD family member 2MAD, mothers against decapentaplegic homolog 2 (Drosophila), SMAD, mothers against DPP homolog 2, Smad2 | |
| This antibody was developed using a synthetic peptide derived from Q15796, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Metastasis, Phospho Specific, Stem Cell Signaling Pathway, Wnt Signaling Pathway | |
| 4087 | |
| Store at 4⪚C in the dark. | |
| IgG2a |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL8472 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction