Learn More
Smad4 Antibody (CL9774) [DyLight 405], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-05507V
Product Code. 30297793
  • 737.00 € / 0.10mL
Description
Smad4 Monoclonal antibody specifically detects Smad4 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Smad4
Monoclonal
DyLight 405
DPC4SMAD, mothers against DPP homolog 4 (Drosophila), hSMAD4, JIP, MAD homolog 4, MADH4mothers against decapentaplegic homolog 4, mothers against decapentaplegic homolog 4, mothers against decapentaplegic, Drosophila, homolog of, 4, SMAD 4, SMAD family member 4MAD, mothers against decapentaplegic homolog 4 (Drosophila), SMAD, mothers against DPP homolog 4, Smad4
This antibody was developed using a recombinant protein derived from Q13485, with the exact immunogen sequence remaining proprietary.
0.1 mL
Signal Transduction, Stem Cell Signaling Pathway
4089
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL9774
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified