missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Smad7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 12 publications
226.00 € - 500.00 €
Specifications
| Applications | Western Blot, Immunohistochemistry |
|---|---|
| Classification | Polyclonal |
| Form | Purified |
| Host Species | Rabbit |
| Target Species | Human |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18787233
|
Novus Biologicals
NBP2-24710SS |
0.025 mg |
226.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18728203
|
Novus Biologicals
NBP2-24710 |
500.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Smad7 Polyclonal specifically detects Smad7 in Human, Mouse, Sheep samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| Western Blot, Immunohistochemistry | |
| Purified | |
| Human | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Polyclonal | |
| Rabbit | |
| IgG |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title