missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Small Intestine Tissue Slides (Malignant Lymphoma)- Paraffin
Shop All Bio Techne ProductsDescription
These slides are paraffin coated to prevent sample oxidization, it is recommended that slides are first de-paraffinized by baking at 60 degrees C for 1 hour in a vertical orientation prior to performing antigen retrieval procedures.
Specifications
Specifications
| Cross Reactivity | Human |
| Type | Small Intestine Tissue Slides (Malignant Lymphoma) |
| Regulatory Status | This product is for research use only and is not approved for use in humans or in clinical diagnosis. |
| Species | Human |
| Tissue Condition | Malignant Lymphoma |
| Product Line | Novus |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?