missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMAP2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
292.00 € - 539.00 €
Specifications
| Antigen | SMAP2 |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18416650
|
Novus Biologicals
NBP1-80737-25ul |
25ul |
292.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18264387
|
Novus Biologicals
NBP1-80737 |
0.1 mL |
539.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
SMAP2 Polyclonal specifically detects SMAP2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Spezifikation
| SMAP2 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 64744 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:ASGMVAPMAMPAGYMGGMQASMMGVPNGMMTTQQAGYMAGMAAMPQTVYGVQPAQQLQWNLTQMTQQMAGMNFYGANGMMNYGQSMSGGNGQAANQTLSPQMWK | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Mouse, Rat, Human | |
| RP1-228H13.3, small ArfGAP2, SMAP1L, SMOC2, stromal membrane-associated GTPase-activating protein 2, Stromal membrane-associated protein 1-like, stromal membrane-associated protein 2 | |
| SMAP2 | |
| IgG | |
| Affinity Purified | |
| Specificity of human SMAP2 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts