missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMARCAD1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00 € - 470.00 €
Specifications
| Antigen | SMARCAD1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18644342
|
Novus Biologicals
NBP2-94750-0.02ml |
0.02 mL |
190.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18627122
|
Novus Biologicals
NBP2-94750-0.1ml |
0.1 mL |
470.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SMARCAD1 Polyclonal antibody specifically detects SMARCAD1 in Human samples. It is validated for Western BlotSpecifications
| SMARCAD1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| DNA Repair | |
| PBS (pH 7.3), 50% glycerol | |
| 56916 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| ATP-dependent helicase 1, DKFZP762K2015, EC 3.6.1, EC 3.6.4.12, HEL1, hHEL1, KIAA1122ETL1DKFZp762K2015, subfamily a, containing DEAD/H box 1, SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily A containing DEAD/H box 1, SWI/SNF-related, matrix-associated actin-dependent regulator of chromatin | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human SMARCAD1 (NP_001121901.1). MNLFNLDRFRFEKRNKIEEAPEATPQPSQPGPSSPISLSAEEENAEGEVSRANTPDSDITEKTEDSSVPETPDNERKASISYFKNQRGIQYIDLSSDSEDVVSPNCSNTVQEKTFNKDTVIIVSEPSEDEESQGLPTMARRNDDISELEDLSELEDLKDAKLQTLKELFPQRSDNDLLKLIESTSTMDGAIAAALLMFGDAGGGPRKRKLSSSSEPYEEDEFNDDQSIKKTRLDHGEESNESAESSSNWEKQESIVLKLQKEFPNFDKQELREVLKEHEWMYTEALESLKVFAEDQDMQY | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title