missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMARCAD1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
369.00 € - 533.00 €
Specifications
| Antigen | SMARCAD1 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18403301
|
Novus Biologicals
NBP1-85023-25ul |
25ul |
369.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18035545
|
Novus Biologicals
NBP1-85023 |
0.1 mL |
533.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SMARCAD1 Polyclonal specifically detects SMARCAD1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
| SMARCAD1 | |
| Polyclonal | |
| Rabbit | |
| DNA Repair | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 56916 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:HGEESNESAESSSNWEKQESIVLKLQKEFPNFDKQELREVLKEHEWMYTEALESLKVFAEDQDMQYVSQSEVPNGKEVSSRSQNYPKNATKTKLKQKFSMKAQNGFNKK | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| ATP-dependent helicase 1, DKFZP762K2015, EC 3.6.1, EC 3.6.4.12, HEL1, hHEL1, KIAA1122ETL1DKFZp762K2015, subfamily a, containing DEAD/H box 1, SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily A containing DEAD/H box 1, SWI/SNF-related, matrix-associated actin-dependent regulator of chromatin | |
| SMARCAD1 | |
| IgG | |
| Affinity Purified | |
| Specificity of human SMARCAD1 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title