missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMC4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97937-50ul
This item is not returnable.
View return policy
Description
SMC4 Polyclonal antibody specifically detects SMC4 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| SMC4 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:200-1:1000 | |
| CAP-C, CAPCSMC4 structural maintenance of chromosomes 4-like 1 (yeast), Chromosome-associated polypeptide C, hCAP-CSMC4 (structural maintenance of chromosomes 4, yeast)-like 1, SMC protein 4, SMC-4, SMC4 structural maintenance of chromosomes 4-like 1, SMC4L1, structural maintenance of chromosomes 4, structural maintenance of chromosomes protein 4, XCAP-C homolog | |
| Produced in rabbits immunized with E. coli-derived Human SMC4 fragment. | |
| 50 μg | |
| Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
| 10051 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction