Learn More
SMCHD1 Antibody (CL4270) [Alexa Fluor™ 750], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-59046AF750
Product Code. 30300449
  • 737.00 € / 0.10mL
Description
SMCHD1 Monoclonal antibody specifically detects SMCHD1 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SMCHD1
Monoclonal
Alexa Fluor 750
DKFZp686O0631, KIAA0650, structural maintenance of chromosomes flexible hinge domain containing 1, structural maintenance of chromosomes flexible hinge domain-containing protein 1
This antibody was developed using a recombinant protein derived from A6NHR9, with the exact immunogen sequence remaining proprietary.
0.1 mL
Protein Kinase
23347
Store at 4⪚C in the dark.
IgG2b
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL4270
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified