missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SMEK2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17299223 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17299223 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17299223 Supplier Invitrogen™ Supplier No. PA5116728

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Samples: Mouse testis Immunogen sequence: YKALESIEYV QTFKGLKTKY EQEKDRQNQK LNSVPSILRS NRFRRDAKAL EEDEEMWFNE DEEEEGKAVV APVEKPKPED DFPDNYEKFM ETKKAKESED KENLPKRTSP GGFKFTFSHS ASAANGTNSK SVVAQIPPAT SNGSSSKTTN LPTSVTATKG SLVGLVDYPD DEEEDEEEES SPRKRPRLGS.

Regulatory subunit of serine/threonine-protein phosphatase 4 (PP4). May regulate the activity of PPP4C at centrosomal microtubule organizing centers. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMEK2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.68 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene PPP4R3B
Gene Accession No. Q5MIZ7, Q922R5
Gene Alias AW011752; AW557776; FLFL2; KIAA1387; mKIAA1387; Pp4r3b; PPP4R3B; protein phosphatase 4 regulatory subunit 3B; protein phosphatase 4, regulatory subunit 3B; PSY2; RGD1309450; Serine/threonine-protein phosphatase 4 regulatory subunit 3B; SMEK homolog 2; SMEK homolog 2, suppressor of mek1; SMEK homolog 2, suppressor of mek1 (Dictyostelium); SMEK2; smk1
Gene Symbols PPP4R3B
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 628-817 of human PPP4R3B (NP_0012697791).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 104570, 360993, 57223
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.