missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SMOC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA550768
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The antibody detects endogenous levels of total SMOC2 protein.
This gene encodes a member of the SPARC family (secreted protein acidic and rich in cysteine/osteonectin/BM-40), which are highly expressed during embryogenesis and wound healing. The gene product is a matricellular protein which promotes matrix assembly and can stimulate endothelial cell proliferation and migration, as well as angiogenic activity. Associated with pulmonary function, this secretory gene product contains a Kazal domain, two thymoglobulin type-1 domains, and two EF-hand calcium-binding domains. The encoded protein may serve as a target for controlling angiogenesis in tumor growth and myocardial ischemia. Alternative splicing results in multiple transcript variants.
Spécification
| SMOC2 | |
| Polyclonal | |
| Unconjugated | |
| SMOC2 | |
| bA270C4A.1; bA37D8.1; dJ421D16.1; DTDP1; MST117; MSTP117; MSTP140; secreted modular calcium-binding protein 2; SMAP2; SMAP-2; SMOC2; SMOC-2; smooth muscle associated protein 2; smooth muscle-associated protein 2; SPARC related modular calcium binding 2; SPARC-related modular calcium-binding protein 2; thyroglobulin type-1 repeat containing protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 64094 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 2.3 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q9H3U7 | |
| SMOC2 | |
| Synthetic peptide corresponding to residues near the C terminal of human SPARC related modular calcium binding 2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu