missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Smoothelin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
280.00 € - 617.00 €
Specifications
| Antigen | Smoothelin |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18749473
|
Novus Biologicals
NBP2-37931 |
0.1 mL |
617.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18661646
|
Novus Biologicals
NBP2-37931-25ul |
25 μL |
280.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Smoothelin Polyclonal specifically detects Smoothelin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Smoothelin | |
| Polyclonal | |
| Rabbit | |
| Human | |
| P53814 | |
| 6525 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: LAGLDEGALRKLLEVTADLAERRRIRSAIRELQRQELEREEEALASKRFRAERQDNKENWLHSQQREAEQRAALARLAGQLESMNDVEELTALLR | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| FLJ35365, FLJ38597, smoothelin, SMSMO | |
| SMTN | |
| IgG | |
| Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title