missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMURF1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
178.00 € - 507.00 €
Specifications
| Antigen | SMURF1 |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18653357
|
Novus Biologicals
NBP2-41297-0.025mg |
0.025 mg |
178.00 €
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18141668
|
Novus Biologicals
NBP2-41297 |
0.1 mg |
507.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
SMURF1 Polyclonal specifically detects SMURF1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Spezifikation
| SMURF1 | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| E3 ubiquitin ligase SMURF1, EC 6.3.2, EC 6.3.2.-, hSMURF1, KIAA1625E3 ubiquitin-protein ligase SMURF1, SMAD specific E3 ubiquitin protein ligase 1, SMAD ubiquitination regulatory factor 1, Smad-specific E3 ubiquitin ligase 1, SMAD-specific E3 ubiquitin-protein ligase 1 | |
| SMURF1 | |
| IgG | |
| Affinity Purified | |
| 78 kDa |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Stem Cell Signaling Pathway | |
| PBS with 0.02% Sodium Azide | |
| 57154 | |
| Antibody was raised against a 16 amino acid peptide near the amino terminus of human SMURF1. The immunogen is located within amino acids 180 - 230 of SMURF1. Amino Acid Squence: RFVESPSQDQRLQAQ | |
| Primary | |
| At least three isoforms of SMURF1 are known to exist this antibody will recognize all three isoforms. SMURF1 is predicted to not cross-react with SMURF2. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts