missing translation for 'onlineSavingsMsg'
Learn More

Smurf1-IN-A01, MedChemExpress

Product Code. 16490687
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16490687 50 mg 50mg
16473046 25 mg 25mg
16425276 100 mg 100mg
16424368 10 mg 10mg
16407087 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16490687 Supplier Medchem Express Supplier No. HY110195
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Smurf1-IN-A01 (A01) is an ubiquitin ligase Smad ubiquitination regulatory factor-1 (Smurf1) inhibitor with a kd of 3.664 nM, which increases BMP-2 responsiveness by inhibiting Smurf1-mediated Smad1/5 degradation.

Smurf1-IN-A01 (A01) is an ubiquitin ligase Smad ubiquitination regulatory factor-1 (Smurf1) inhibitor with a kd of 3.664 nM, which increases BMP-2 responsiveness by inhibiting Smurf1-mediated Smad1/5 degradation.

Specifications

Chemical Name or Material Smurf1-IN-A01
CAS 1007647-73-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C22H20ClF3N4O3S
Quantity 50 mg
Synonym A01
Solubility Information DMSO : 62.5 mg/mL (121.85 mM; Need ultrasonic)
SMILES O=S(C1=CC(C(F)(F)F)=C(Cl)C=C1)(N2CCN(C(C3=CC=C(N4C(C)=CC=N4)C=C3)=O)CC2)=O
Molecular Weight (g/mol) 512.93
Formula Weight 512.93
Percent Purity 99.58%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.