missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SNAP29 Recombinant Rabbit Monoclonal Antibody (HL3232)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556877
This item is not returnable.
View return policy
Description
SNAP29 Recombinant Monoclonal Antibody for Western Blot
SNAP29 encodes a product that is a member of the SNAP25 gene family, encodes a protein involved in multiple membrane trafficking steps. Two other members of this gene family, SNAP23 and SNAP25, encode proteins that bind a syntaxin protein and mediate synaptic vesicle membrane docking and fusion to the plasma membrane. The protein encoded by this gene binds tightly to multiple syntaxins and is localized to intracellular membrane structures rather than to the plasma membrane. While the protein is mostly membrane-bound, a significant fraction of it is found free in the cytoplasm. Use of multiple polyadenylation sites has been noted for this gene.
Specifications
| SNAP29 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| O95721 | |
| Snap29 | |
| Recombinant fragment of human SNAP29. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| HL3232 | |
| Unconjugated | |
| Snap29 | |
| 1300018G05Rik; AI891940; AU020222; BB131856; CEDNIK; fc84f09; FLJ21051; golgi SNARE of 32 kDa; Gs32; im:7145101; LOC553460 protein; si:dkey-178e17.4; SNAP29; SNAP-29; Soluble 29 kDa NSF attachment protein; synaptosomal-associated protein 29; synaptosomal-associated protein 29kD; synaptosomal-associated protein, 29kD; synaptosomal-associated protein, 29kDa; synaptosome associated protein 29; synaptosome associated protein 29kDa; vesicle-membrane fusion protein SNAP-29; wu:fc84f09 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 9342 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction