missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNAP45 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00 € - 470.00 €
Specifications
| Antigen | SNAP45 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18609840
|
Novus Biologicals
NBP2-93703-0.02ml |
0.02 mL |
190.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18697060
|
Novus Biologicals
NBP2-93703-0.1ml |
0.1 mL |
470.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SNAP45 Polyclonal antibody specifically detects SNAP45 in Human, Mouse samples. It is validated for Western BlotSpecifications
| SNAP45 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| Proximal sequence element-binding transcription factor subunit delta, PSE-binding factor subunit delta, PTF subunit delta, PTFdelta, small nuclear RNA activating complex, polypeptide 2, 45kD, small nuclear RNA activating complex, polypeptide 2, 45kDa, Small nuclear RNA-activating complex polypeptide 2, SNAP45snRNA-activating protein complex 45 kDa subunit, SNAPc 45 kDa subunit, SNAPc subunit 2, snRNA-activating protein complex subunit 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 20-100 of human SNAPC2 (NP_003074.1). GPATWSAREKRQLVRLLQARQGQPEPDATELARELRGRSEAEIRVFLQQLKGRVAREAIQKVHPGGLQGPRRREAQPPAPI | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500 - 1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 6618 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title