missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNF5 Antibody (CL13973) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-43860
This item is not returnable.
View return policy
Description
SNF5 Monoclonal antibody specifically detects SNF5 in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry
Specifications
| SNF5 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 μg/mL, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| BAF47, BAF47hSNF5, BRG1-associated factor 47, hSNFS, Ini1, INI1RTPS1, Integrase interactor 1 protein, malignant rhabdoid tumor suppressor, RDTSNF5 homolog, Sfh1p, SNF5, SNF5L1, Snr1, subfamily b, member 1, sucrose nonfermenting, yeast, homolog-like 1, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily B member 1 | |
| This antibody was generated using a recombinant protein sequence of Q12824, with the exact immunogen sequence remaining proprietary. | |
| 100 μL | |
| Cell Cycle and Replication | |
| 6598 | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry | |
| CL13973 | |
| Unconjugated | |
| Lyophilized in 5% Trehalose. | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyophilized antibody at 12,000 x g for 20 seconds. Reconstitute by adding sterile, distilled water to achieve a final antibody concentration of 1mg/mL. | |
| Store at 4⪚C short term. Aliquot and store at -20⪚C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction