missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNF5 Antibody (SMARCB1/8851R), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-23874-100ug
This item is not returnable.
View return policy
Description
SNF5 Monoclonal antibody specifically detects SNF5 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| SNF5 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| BAF47, BAF47hSNF5, BRG1-associated factor 47, hSNFS, Ini1, INI1RTPS1, Integrase interactor 1 protein, malignant rhabdoid tumor suppressor, RDTSNF5 homolog, Sfh1p, SNF5, SNF5L1, Snr1, subfamily b, member 1, sucrose nonfermenting, yeast, homolog-like 1, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily B member 1 | |
| Recombinant fragment (around aa1-200) of human SNF5 protein (exact sequence is proprietary) | |
| 100 μg | |
| Cell Cycle and Replication | |
| 6598 | |
| Store at 4°C. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| SMARCB1/8851R | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction