missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SNRNP27 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15734871 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15734871 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15734871 Supplier Invitrogen™ Supplier No. PA556970

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: TSPSPSRLKE RRDEEKKETK ETKSKERQIT EEDLEGKTEE EIEMMKLMGF ASFDSTKGKK VDGSVNAYAI NVSQKRKYRQ YMNRKGGFNR Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

May play a role in mRNA splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNRNP27
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SNRNP27
Gene Accession No. Q8WVK2
Gene Alias 2610209M04Rik; 27K; AI449063; AU022368; Nucleic acid-binding protein RY-1; putative nucleic acid binding protein RY-1; RY1; small nuclear ribonucleoprotein 27 (U4/U6.U5); small nuclear ribonucleoprotein 27kDa (U4/U6.U5); small nuclear ribonucleoprotein U4/U6.U5 subunit 27; small nuclear ribonucleoprotein, U4/U6.U5 27kDa subunit; Snrnp27; U4/U6.U5 small nuclear ribonucleoprotein 27 kDa protein; U4/U6.U5 snRNP 27 kDa protein; U4/U6.U5 tri-snRNP-associated 27 kDa protein; U4/U6.U5 tri-snRNP-associated protein 3; U4/U6.U5-27K; U4/U6U5-27K
Gene Symbols SNRNP27
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SNRNP27. Recombinant protein control fragment (Product #RP-96065).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11017
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.