missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNTA1 Monoclonal Antibody (OTI2D5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502041S
This item is not returnable.
View return policy
Description
SNTA1 Monoclonal antibody specifically detects SNTA1 in Canine, Human, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| SNTA1 | |
| Monoclonal | |
| 0.28 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q13424 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 362242, 485837, 6640 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Flow Cytometry, Immunocytochemistry, Western Blot | |
| OTI2D5 | |
| Unconjugated | |
| SNTA1 | |
| 59 kDa dystrophin-associated protein A1 acidic component 1; 59-1 DAP; acidic alpha 1 syntrophin; alpha-1-syntrophin; alpha1-syntrophin; alpha-syntrophin; AW228934; dJ1187J4.5; dystrophin-associated protein A1, 59kDa, acidic component; LQT12; pro-TGF-alpha cytoplasmic domain-interacting protein 1; RP23-192L3.1; SNT1; SNTA1; syntrophin alpha 1; syntrophin, acidic 1; syntrophin, alpha 1; syntrophin, alpha 1 (dystrophin-associated protein A1, 59kDa, acidic component); syntrophin-1; TACIP1; Unknown (protein for MGC:142519) | |
| Full length human recombinant protein of human SNTA1 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Canine, Human, Rat | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction